Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPS3F
Entrez Gene ID
839382
Gene Name
sucrose phosphate synthase 3F
Gene Aliases
AT1G04920, ATSPS3F, F13M7.9, F13M7_9, sucrose phosphate synthase 3F
Chromosome Location
Chr.1: 1391434 - 1395953 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100370.3 NP_171984.2 4 - 5 1488 76
GenBank mRNA AY078949.1 - 4 - 5 1399 76
BT002210.1 - 4 - 5 1291 76
KJ139031.1 - 4 - 5 1291 76

View Full Product Details