Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CALS1
Entrez Gene ID
837059
Gene Name
callose synthase 1
Gene Aliases
AT1G05570, ATGSL06, ATGSL6, F3F20.1, F3F20_1, GLUCAN SYNTHASE-LIKE 6, GSL06, GSL6, callose synthase 1
Chromosome Location
Chr.1: 1647575 - 1659611 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197984.1 NP_001184913.1 21 - 22 2255 111
NM_001331555.1 NP_001322482.1 20 - 21 2469 111
NM_001331556.1 NP_001322483.1 19 - 20 2303 111
NM_100436.5 NP_563743.2 20 - 21 2552 111

View Full Product Details