Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLE3
Entrez Gene ID
837132
Gene Name
CLAVATA3/ESR-RELATED 3
Gene Aliases
AT1G06225, CLAVATA3/ESR-RELATED 3
Chromosome Location
Chr.1: 1905918 - 1906298 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100504.2 NP_563763.1 1 - 1 248 63
GenBank mRNA DQ446232.1 - 1 - 1 191 63

View Full Product Details