Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CalS7
Entrez Gene ID
837160
Gene Name
glucan synthase-like 7
Gene Aliases
AT1G06490, ATGSL07, Arabidopsis thaliana glucan synthase-like 7, F12K11.17, F12K11_17, GSL07, GSL7, atgsl7, callose synthase 7, glucan synthase-like 7, gsl07
Chromosome Location
Chr.1: 1978378 - 1989409 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331632.1 NP_001322002.1 23 - 24 3151 142
NM_100528.3 NP_172136.2 23 - 24 2809 142
GenBank mRNA HM049631.1 - 23 - 24 2767 142

View Full Product Details