Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCP4;1
Entrez Gene ID
819082
Gene Name
cyclin p4;1
Gene Aliases
AT2G44740, F16B22.23, cyclin p4;1
Chromosome Location
Chr.2: 18441828 - 18443405 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130038.4 NP_182002.1 1 - 2 436 71
GenBank mRNA AK229380.1 - 1 - 2 427 71
BT021978.1 - 1 - 2 428 71
BT023473.1 - 1 - 2 335 71

View Full Product Details