Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PVA12
Entrez Gene ID
819122
Gene Name
plant VAP homolog 12
Gene Aliases
AT2G45140, T14P1.5, plant VAP homolog 12
Chromosome Location
Chr.2: 18610806 - 18613302 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130077.3 NP_182039.1 6 - 7 752 68
GenBank mRNA AY088399.1 - 6 - 7 691 68
AY093014.1 - 6 - 7 752 68
BT006283.1 - 6 - 7 529 68

View Full Product Details