Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AFO
Entrez Gene ID
819127
Gene Name
Plant-specific transcription factor YABBY family protein
Gene Aliases
ABNORMAL FLORAL ORGANS, ABNORMAL FLORAL ORGANS PROTEIN, AT2G45190, F4L23.30, FIL, FILAMENTOUS FLOWER, YAB1, YABBY1
Chromosome Location
Chr.2: 18628252 - 18630779 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337117.1 NP_001323472.1 5 - 6 611 83
NM_130082.4 NP_566037.1 6 - 7 841 83
GenBank mRNA AF074948.1 - 6 - 7 614 83
AF087015.1 - 6 - 7 726 83
AF136538.1 - 6 - 7 726 83
BT026409.1 - 6 - 7 614 83

View Full Product Details