Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAM7
Entrez Gene ID
819196
Gene Name
beta-amylase 7
Gene Aliases
AT2G45880, BETA-AMYLASE 4, BMY4, F4I18.14, beta-amylase 7
Chromosome Location
Chr.2: 18878518 - 18882995 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130151.7 NP_182112.2 7 - 8 2369 83

View Full Product Details