Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AIB
- Entrez Gene ID
-
819262
- Gene Name
- ABA-inducible BHLH-type transcription factor
- Gene Aliases
-
ABA-inducible BHLH-type transcription factor, AT2G46510, ATAIB, F11C10.32, JA-associated MYC2-like 1, JAM1
- Chromosome Location
-
Chr.2: 19090953 - 19093373 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_130216.3 |
NP_566078.1 |
1 - 1 |
2110 |
93 |
View Full Product Details