Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LIG1
Entrez Gene ID
837333
Gene Name
DNA ligase 1
Gene Aliases
AT1G08130, ATLIG1, DNA ligase 1, T6D22.23
Chromosome Location
Chr.1: 2542663 - 2547946 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100689.5 NP_172293.2 7 - 8 1666 100
GenBank mRNA AK117238.1 - 7 - 8 1674 100
BT005964.1 - 7 - 8 1535 100

View Full Product Details