Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSAO
Entrez Gene ID
837358
Gene Name
photosystem I subunit O
Gene Aliases
AT1G08380, PSI-O, T27G7.25, T27G7_25, photosystem I subunit O
Chromosome Location
Chr.1: 2640803 - 2641934 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100711.4 NP_563815.1 1 - 2 267 67
GenBank mRNA AJ493060.1 - 1 - 2 103 67
AY050960.1 - 1 - 2 96 67
AY087368.1 - 1 - 2 161 67
AY091345.1 - 1 - 2 72 67

View Full Product Details