Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALB1
Entrez Gene ID
837374
Gene Name
ALBINA 1
Gene Aliases
ALB-1V, ALBINA 1, AT1G08520, CHLD, PDE166, PIGMENT DEFECTIVE EMBRYO 166, T27G7.20, T27G7_20, V157
Chromosome Location
Chr.1: 2696381 - 2701022 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100725.4 NP_563821.2 2 - 3 607 102
GenBank mRNA AK226535.1 - 2 - 3 492 102
AX765812.1 - 2 - 3 450 102
AY063821.1 - 2 - 3 508 102
AY091402.1 - 2 - 3 450 102

View Full Product Details