Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACHT4
Entrez Gene ID
837379
Gene Name
atypical CYS HIS rich thioredoxin 4
Gene Aliases
AT1G08570, atypical CYS HIS rich thioredoxin 4, atypical CYS HIS rich thioredoxin 4
Chromosome Location
Chr.1: 2712694 - 2714833 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084022.1 NP_001077491.1 2 - 3 311 69
NM_001123776.1 NP_001117248.1 2 - 3 328 69
NM_001123777.1 NP_001117249.1 2 - 3 256 69
NM_100730.4 NP_172333.1 1 - 2 602 69
GenBank mRNA AF144387.1 - 1 - 2 347 69
AK318661.1 - 2 - 3 256 69
AY034938.1 - 1 - 2 369 69
AY063115.1 - 1 - 2 237 69

View Full Product Details