Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BON3
Entrez Gene ID
837408
Gene Name
Calcium-dependent phospholipid-binding Copine family protein
Gene Aliases
AT1G08860, BONZAI 3, F7G19.25, F7G19_25
Chromosome Location
Chr.1: 2841969 - 2846350 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100759.4 NP_172362.3 4 - 5 595 122
GenBank mRNA AY741136.1 - 4 - 5 411 122

View Full Product Details