Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERD6
Entrez Gene ID
837414
Gene Name
Major facilitator superfamily protein
Gene Aliases
AT1G08930, EARLY RESPONSE TO DEHYDRATION 6, F7G19.19, F7G19_19
Chromosome Location
Chr.1: 2873300 - 2877270 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035929.1 NP_001031006.1 11 - 12 1102 109
NM_100765.5 NP_563830.1 11 - 12 1267 109
GenBank mRNA AJ249967.1 - 11 - 12 1059 109
BT008661.1 - 11 - 12 1077 109
D89051.1 - 11 - 12 1002 109

View Full Product Details