Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERD6
Entrez Gene ID
837414
Gene Name
Major facilitator superfamily protein
Gene Aliases
AT1G08930, EARLY RESPONSE TO DEHYDRATION 6, F7G19.19, F7G19_19
Chromosome Location
Chr.1: 2873300 - 2877270 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035929.1 NP_001031006.1 1 - 2 364 58
NM_100765.5 NP_563830.1 1 - 2 529 58
GenBank mRNA AJ249967.1 - 1 - 2 321 58
BT008661.1 - 1 - 2 339 58
D89051.1 - 1 - 2 264 58

View Full Product Details