Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ADL6
- Entrez Gene ID
-
837568
- Gene Name
- dynamin-like protein 6
- Gene Aliases
-
AT1G10290, DRP2A, DYNAMIN-RELATED PROTEIN 2A, F14N23.17, F14N23_17, dynamin-like protein 6
- Chromosome Location
-
Chr.1: 3370402 - 3377463 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100903.4 |
NP_172500.1 |
21 - 22 |
2874 |
69 |
View Full Product Details