Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEX1
Entrez Gene ID
837619
Gene Name
Pyruvate phosphate dikinase, PEP/pyruvate binding domain-containing protein
Gene Aliases
AT1G10760, GWD, GWD1, SOP, SOP1, STARCH EXCESS 1, T16B5.10, T16B5_10
Chromosome Location
Chr.1: 3580830 - 3590928 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331926.1 NP_001318975.1 28 - 29 3861 105
NM_001331927.1 NP_001320611.1 27 - 28 3710 105
NM_100952.4 NP_563877.1 29 - 30 3695 105
GenBank mRNA AF312027.1 - 26 - 27 3401 105

View Full Product Details