Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RECQ4A
Entrez Gene ID
837636
Gene Name
DNA helicase (RECQl4A)
Gene Aliases
AT1G10930, ATRECQ4A, ATSGS1, T19D16.15, T19D16_15
Chromosome Location
Chr.1: 3647876 - 3655685 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100968.3 NP_172562.2 5 - 6 958 115
GenBank mRNA AY120761.1 - 5 - 6 856 115
BT010133.1 - 4 - 5 652 115

View Full Product Details