Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl31
Entrez Gene ID
837649
Gene Name
serine carboxypeptidase-like 31
Gene Aliases
AT1G11080, T19D16.4, T19D16_4, serine carboxypeptidase-like 31
Chromosome Location
Chr.1: 3694328 - 3698013 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198028.1 NP_001184957.1 4 - 5 649 82
NM_100981.4 NP_172575.2 4 - 5 854 82
GenBank mRNA BT030381.1 - 4 - 5 600 82

View Full Product Details