Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HAG2
- Entrez Gene ID
-
835775
- Gene Name
- histone acetyltransferase of the GNAT family 2
- Gene Aliases
-
AT5G56740, HAC07, HAC7, HAG02, MIK19.19, MIK19_19, histone acetyltransferase of the GNAT family 2
- Chromosome Location
-
Chr.5: 22952767 - 22955739 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125057.4 |
NP_200485.1 |
1 - 2 |
238 |
89 |
GenBank mRNA |
BT025320.1 |
- |
1 - 2 |
76 |
89 |
View Full Product Details