Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ORP4C
Entrez Gene ID
835830
Gene Name
OSBP(oxysterol binding protein)-related protein 4C
Gene Aliases
AT5G57240, MJB24.5, MJB24_5, OSBP(oxysterol binding protein)-related protein 4C
Chromosome Location
Chr.5: 23192872 - 23195476 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037011.2 NP_001032088.1 4 - 5 659 101
NM_001345250.1 NP_001329986.1 3 - 4 1066 101
NM_001345251.1 NP_001329984.1 4 - 5 857 101
NM_001345252.1 NP_001329985.1 4 - 5 857 101
NM_125106.3 NP_200534.1 3 - 4 1066 101
GenBank mRNA AK117264.1 - 3 - 4 617 101

View Full Product Details