Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VDAC4
Entrez Gene ID
835853
Gene Name
voltage dependent anion channel 4
Gene Aliases
ARABIDOPSIS THALIANA VOLTAGE DEPENDENT ANION CHANNEL 4, AT5G57490, ATVDAC4, MUA2.6, MUA2_6, voltage dependent anion channel 4
Chromosome Location
Chr.5: 23283617 - 23286005 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125130.3 NP_200557.1 5 - 6 643 89
GenBank mRNA AY034928.1 - 5 - 6 497 89
AY059112.1 - 4 - 5 311 89
AY086306.1 - 5 - 6 572 89

View Full Product Details