Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRPC1
Entrez Gene ID
836126
Gene Name
nuclear RNA polymerase C1
Gene Aliases
AT5G60040, MGO3.13, MGO3_13, nuclear RNA polymerase C1
Chromosome Location
Chr.5: 24173259 - 24183641 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203644.1 NP_001190573.1 10 - 11 1543 70
NM_125397.3 NP_200812.2 10 - 11 1409 70

View Full Product Details