Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OXA1
Entrez Gene ID
836325
Gene Name
inner membrane OXA1-like protein
Gene Aliases
ARABIDOPSIS THALIANA HOMOLOG OF YEAST OXIDASE ASSEMBLY 1 (OXA1), AT5G62050, ATOXA1, HOMOLOG OF YEAST OXIDASE ASSEMBLY 1 (OXA1) IN ARABIDOPSIS THALIANA, MTG10.19, MTG10_19, OXA1AT, homolog of yeast oxidase assembly 1 (OXA1)
Chromosome Location
Chr.5: 24923465 - 24927314 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125598.4 NP_201011.1 4 - 5 729 63
GenBank mRNA AY140006.1 - 4 - 5 728 63
BT006574.1 - 3 - 4 594 63
Y11871.1 - - 708 63

View Full Product Details