Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALDH12A1
Entrez Gene ID
836373
Gene Name
aldehyde dehydrogenase 12A1
Gene Aliases
ARABIDOPSIS THALIANA DELTA1-PYRROLINE-5-CARBOXYLATE DEHYDROGENASE, AT5G62530, ATP5CDH, DELTA1-PYROLINE-5-CARBOXYLATE DEHYDROGENASE, DELTA1-PYRROLINE-5-CARBOXYLATE DEHYDROGENASE, K19B1.14, K19B1_14, P5CDH, aldehyde dehydrogenase 12A1
Chromosome Location
Chr.5: 25099003 - 25103312 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125647.4 NP_568955.1 5 - 6 555 69
GenBank mRNA AY065072.1 - 5 - 6 452 69
BT000746.1 - 5 - 6 485 69
BT010391.1 - 5 - 6 402 69

View Full Product Details