Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAB8
Entrez Gene ID
841399
Gene Name
poly(A) binding protein 8
Gene Aliases
AT1G49760, F14J22.3, F14J22_3, PABP8, POLY(A) BINDING PROTEIN 8, poly(A) binding protein 8
Chromosome Location
Chr.1: 18416474 - 18420111 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198255.1 NP_001185184.1 2 - 3 680 76
NM_103863.3 NP_564554.1 2 - 3 679 76
GenBank mRNA AF370517.1 - 2 - 3 674 76
AY064622.1 - 2 - 3 414 76

View Full Product Details