Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FU
Entrez Gene ID
841446
Gene Name
kinase family with ARM repeat domain-containing protein
Gene Aliases
AT1G50240, FUSED
Chromosome Location
Chr.1: 18606511 - 18614500 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333427.1 NP_001319189.1 10 - 11 1052 112
NM_001333428.1 NP_001321239.1 10 - 11 985 112

View Full Product Details