Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HKL1
Entrez Gene ID
841468
Gene Name
hexokinase-like 1
Gene Aliases
AT1G50460, ATHKL1, F11F12.18, F11F12_18, hexokinase-like 1
Chromosome Location
Chr.1: 18693644 - 18697709 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333438.1 NP_001322009.1 3 - 4 991 121
NM_001333439.1 NP_001322008.1 3 - 4 548 121
NM_103929.4 NP_175463.1 3 - 4 991 121
GenBank mRNA AY074314.1 - 3 - 4 915 121
AY096416.1 - 3 - 4 604 121

View Full Product Details