Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCPL19
Entrez Gene ID
830823
Gene Name
serine carboxypeptidase-like 19
Gene Aliases
AT5G09640, F17I14.170, F17I14_170, SINAPOYLGLUCOSE ACCUMULATOR 2, SNG2, serine carboxypeptidase-like 19
Chromosome Location
Chr.5: 2988314 - 2991156 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121001.2 NP_568215.2 4 - 5 529 76
GenBank mRNA AY063833.1 - 4 - 5 529 76
AY091309.1 - 4 - 5 470 76

View Full Product Details