Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PMDH2
Entrez Gene ID
830825
Gene Name
peroxisomal NAD-malate dehydrogenase 2
Gene Aliases
AT5G09660, F17I14.150, F17I14_150, peroxisomal NAD-malate dehydrogenase 2
Chromosome Location
Chr.5: 2993411 - 2995784 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036783.1 NP_001031860.1 3 - 4 763 64
NM_001085087.2 NP_001078556.1 3 - 4 968 64
NM_001125727.1 NP_001119199.1 4 - 5 812 64
NM_001343054.1 NP_001330735.1 4 - 5 809 64
NM_121003.4 NP_196528.1 4 - 5 921 64
GenBank mRNA AF428373.1 - 4 - 5 714 64
AJ131206.1 - 4 - 5 718 64
AY037252.1 - 4 - 5 739 64
AY077653.1 - 4 - 5 688 64

View Full Product Details