Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CESA5
- Entrez Gene ID
-
830847
- Gene Name
- cellulose synthase 5
- Gene Aliases
-
AT5G09870, MUCILAGE-MODIFIED 3, MUM3, MYH9.8, MYH9_8, cellulose synthase 5
- Chromosome Location
-
Chr.5: 3072870 - 3078416 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121024.3 |
NP_196549.1 |
1 - 2 |
564 |
138 |
View Full Product Details