Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALF4
Entrez Gene ID
830970
Gene Name
aberrant root formation protein
Gene Aliases
ABERRANT LATERAL ROOT FORMATION 4, AT5G11030, T5K6.20, T5K6_20
Chromosome Location
Chr.5: 3490788 - 3495173 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161232.2 NP_001154704.2 6 - 7 818 85
NM_001161233.1 NP_001154705.1 6 - 7 777 85
NM_001343149.1 NP_001332043.1 6 - 7 780 85
NM_001343150.1 NP_001332042.1 6 - 7 780 85
NM_001343151.1 NP_001332040.1 6 - 7 780 85
NM_001343152.1 NP_001332041.1 6 - 7 777 85
NM_001343153.1 NP_001332045.1 6 - 7 777 85
NM_001343154.1 NP_001332044.1 6 - 7 777 85
NM_121141.5 NP_196664.4 6 - 7 818 85

View Full Product Details