Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC10
Entrez Gene ID
831112
Gene Name
exocyst complex component sec10
Gene Aliases
AT5G12370, exocyst complex component sec10
Chromosome Location
Chr.5: 4002742 - 4008804 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036794.2 NP_001031871.1 6 - 7 797 84
NM_001036795.2 NP_001031872.1 6 - 7 668 84
NM_121275.4 NP_568270.3 6 - 7 669 84
GenBank mRNA AK222187.1 - 6 - 7 774 84

View Full Product Details