Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC10
Entrez Gene ID
831112
Gene Name
exocyst complex component sec10
Gene Aliases
AT5G12370, exocyst complex component sec10
Chromosome Location
Chr.5: 4002742 - 4008804 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036794.2 NP_001031871.1 7 - 8 901 68
NM_001036795.2 NP_001031872.1 7 - 8 772 68
NM_121275.4 NP_568270.3 7 - 8 773 68
GenBank mRNA AK222187.1 - 7 - 8 878 68

View Full Product Details