Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PKL
Entrez Gene ID
817055
Gene Name
chromatin remodeling factor CHD3 (PICKLE)
Gene Aliases
AT2G25170, AT2G25170.1, CHD3, CHD3 PROTEIN, CHR6, CKH2, CYTOKININ-HYPERSENSITIVE 2, ENHANCED PHOTOMORPHOGENIC 1, EPP1, GYM, GYMNOS, PICKLE, SSL2, SUPPRESSOR OF SLR 2
Chromosome Location
Chr.2: 10713807 - 10724106 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335976.1 NP_001324292.1 14 - 15 1815 66
NM_001335977.1 NP_001324293.1 14 - 15 1842 66
NM_001335978.1 NP_001324291.1 13 - 14 1764 66
NM_128074.4 NP_565587.1 14 - 15 1788 66
GenBank mRNA AF185577.1 - 14 - 15 1788 66
AF185578.1 - 13 - 14 1768 66

View Full Product Details