Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AUR2
Entrez Gene ID
817129
Gene Name
ataurora2
Gene Aliases
AT2G25880, AtAUR2, F17H15.9, F17H15_9, ataurora2
Chromosome Location
Chr.2: 11034652 - 11038191 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001124914.1 NP_001118386.1 3 - 4 364 79
NM_001336027.1 NP_001325078.1 4 - 5 1559 79
NM_001336028.1 NP_001325077.1 5 - 6 1415 79
NM_128148.4 NP_180159.2 3 - 4 401 79
GenBank mRNA AK175188.1 - 3 - 4 365 79

View Full Product Details