Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIL2
Entrez Gene ID
817254
Gene Name
DNA polymerase epsilon catalytic subunit
Gene Aliases
AT2G27120, POL2B, T20P8.17, T20P8_17, TILTED 2
Chromosome Location
Chr.2: 11581214 - 11594397 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336102.1 NP_001325161.1 43 - 44 5473 79
NM_001336103.1 NP_001325160.1 43 - 44 5467 79
NM_128270.3 NP_180280.2 43 - 44 5464 79

View Full Product Details