Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FUC1
Entrez Gene ID
817355
Gene Name
alpha-L-fucosidase 1
Gene Aliases
AT2G28100, ATFUC1, F24D13.11, F24D13_11, alpha-L-fucosidase 1
Chromosome Location
Chr.2: 11974663 - 11976600 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128370.3 NP_180377.2 1 - 2 685 125

View Full Product Details