Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXO70H8
Entrez Gene ID
817414
Gene Name
exocyst subunit exo70 family protein H8
Gene Aliases
AT2G28650, ATEXO70H8, T8O18.6, T8O18_6, exocyst subunit exo70 family protein H8
Chromosome Location
Chr.2: 12289152 - 12291092 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128426.3 NP_180433.1 1 - 1 1799 110
GenBank mRNA AK118418.2 - 1 - 1 1752 110

View Full Product Details