Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSTU1
Entrez Gene ID
817498
Gene Name
glutathione S-transferase TAU 1
Gene Aliases
AT2G29490, ATGSTU1, F16P2.13, F16P2_13, GLUTATHIONE S-TRANSFERASE 19, GST19, glutathione S-transferase TAU 1
Chromosome Location
Chr.2: 12631341 - 12632538 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128503.3 NP_180510.1 1 - 2 494 88
GenBank mRNA AF288183.1 - 1 - 2 316 88
AF428387.1 - 1 - 2 413 88
BT010162.1 - 1 - 2 316 88

View Full Product Details