Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JOSL
Entrez Gene ID
817514
Gene Name
JOSEPHIN-like protein
Gene Aliases
AT2G29640, JOSEPHIN-like protein, T27A16.26, T27A16_26
Chromosome Location
Chr.2: 12671206 - 12673334 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128518.2 NP_180525.1 2 - 3 509 99

View Full Product Details