Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASA2
Entrez Gene ID
817519
Gene Name
anthranilate synthase 2
Gene Aliases
AT2G29690, ATHANSYNAB, T27A16.21, T27A16_21, anthranilate synthase 2
Chromosome Location
Chr.2: 12693734 - 12697091 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336222.1 NP_001324956.1 2 - 3 536 65
NM_128523.5 NP_180530.1 2 - 3 581 65

View Full Product Details