Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCG31
- Entrez Gene ID
-
817544
- Gene Name
- pleiotropic drug resistance 3
- Gene Aliases
-
AT2G29940, ATP-binding cassette G31, ATPDR3, F23F1.14, F23F1_14, PDR3, PLEIOTROPIC DRUG RESISTANCE 3, pleiotropic drug resistance 3
- Chromosome Location
-
Chr.2: 12759973 - 12766794 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128548.4 |
NP_180555.2 |
14 - 15 |
2685 |
168 |
View Full Product Details