Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP7
Entrez Gene ID
821682
Gene Name
ubiquitin carboxyl-terminal hydrolase
Gene Aliases
AT3G21280, ubiquitin-specific protease 7
Chromosome Location
Chr.3: 7477930 - 7482125 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338521.1 NP_001325430.1 6 - 7 585 85
NM_113023.4 NP_566680.2 7 - 8 838 85

View Full Product Details