Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OASA2
Entrez Gene ID
821817
Gene Name
O-acetylserine (thiol) lyase (OAS-TL)
Gene Aliases
AT3G22460, O-acetylserine (thiol) lyase (OAS-TL) isoform A2
Chromosome Location
Chr.3: 7963855 - 7965907 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338588.1 NP_001326025.1 1 - 2 46 99
GenBank mRNA BT031364.1 - 1 - 2 46 99

View Full Product Details