Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMT3
Entrez Gene ID
821845
Gene Name
homocysteine S-methyltransferase 3
Gene Aliases
AT3G22740, homocysteine S-methyltransferase 3
Chromosome Location
Chr.3: 8032917 - 8036005 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113173.3 NP_566715.1 5 - 6 1089 76
GenBank mRNA AF297394.1 - 5 - 6 794 76
AK118021.1 - 5 - 6 889 76
BT005318.1 - 5 - 6 794 76

View Full Product Details