Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYN
Entrez Gene ID
821930
Gene Name
cyanase
Gene Aliases
AT3G23490, CYANASE, cyanase
Chromosome Location
Chr.3: 8422649 - 8424652 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338643.1 NP_001327887.1 1 - 2 507 123
NM_001338644.1 NP_001327888.1 1 - 2 507 123
NM_113252.4 NP_188991.1 1 - 2 507 123
GenBank mRNA AB004568.1 - 1 - 2 510 123
AF410298.1 - 1 - 2 285 123
AY093708.1 - 1 - 2 270 123

View Full Product Details