Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FMA
Entrez Gene ID
822000
Gene Name
basic helix-loop-helix (bHLH) DNA-binding superfamily protein
Gene Aliases
AT3G24140, FAMA
Chromosome Location
Chr.3: 8715118 - 8718211 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338669.1 NP_001326090.1 3 - 4 883 76
NM_113319.5 NP_189056.2 3 - 4 883 76
GenBank mRNA AK221324.1 - 3 - 4 884 76
BT028961.1 - 2 - 3 688 76

View Full Product Details