Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHUP1
Entrez Gene ID
822157
Gene Name
Hydroxyproline-rich glycoprotein family protein
Gene Aliases
AT3G25690, Arabidopsis thaliana CHLOROPLAST UNUSUAL POSITIONING 1, AtCHUP1, CHLOROPLAST UNUSUAL POSITIONING 1
Chromosome Location
Chr.3: 9352444 - 9357953 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203045.1 NP_001189974.1 5 - 6 2524 130
NM_001203046.1 NP_001189975.1 5 - 6 2101 130
NM_001338761.1 NP_001327702.1 6 - 7 2709 130
NM_001338762.1 NP_001327703.1 6 - 7 2548 130
NM_001338763.1 NP_001327701.1 5 - 6 3524 130
NM_113468.5 NP_189197.2 4 - 5 3107 130
GenBank mRNA AB087408.1 - 4 - 5 2539 130

View Full Product Details